Assay Details
Target Gene Details
Entrez Gene ID: | 7461 |
Gene Name: | CAP-Gly domain containing linker protein 2 |
Gene Aliases: |
CLIP, CLIP-115, CYLN2, WBSCR3, WBSCR4, WSCR3, WSCR4 |
Location: |
Chr.7:74289437-74405943 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLIP2 | NM_003388.4 | NP_003379.3 | ||
| NM_032421.2 | NP_115797.1 | |||
| AB006629.2 | ||||
| BC150332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3613698 | Chr.7:74090462 - 74398730 on Build GRCh38 | Loss |
|
| nsv831031 | Chr.7:74282956 - 74405915 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map