Assay Details
Target Gene Details
Entrez Gene ID: | 101929580 |
Gene Name: | general transcription factor IIi pseudogene 23 |
Gene Aliases: |
- |
Location: |
Chr.7:66847450-66884273 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2IP23 | NR_135738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3613520 | Chr.7:66748771 - 66896312 on Build GRCh38 | Gain |
|
| nsv981514 | Chr.7:66845152 - 66901253 on Build GRCh38 | Duplication |
|
| esv3891136 | Chr.7:66809968 - 67401188 on Build GRCh38 | Gain |
|

Genomic Map