Assay Details
Target Gene Details
Entrez Gene ID: | 9863 |
Gene Name: | membrane associated guanylate kinase, WW and PDZ domain containing 2 |
Gene Aliases: |
ACVRIP1, AIP-1, AIP1, ARIP1, MAGI-2, SSCAM |
Location: |
Chr.7:78017055-79453805 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGI2 | NM_001301128.1 | NP_001288057.1 | ||
| NM_012301.3 | NP_036433.2 | |||
| XM_011516718.1 | XP_011515020.1 | |||
| XM_017012840.1 | XP_016868329.1 | |||
| XM_017012841.1 | XP_016868330.1 | |||
| XM_017012842.1 | XP_016868331.1 | |||
| XM_017012843.1 | XP_016868332.1 | |||
| XM_017012844.1 | XP_016868333.1 | |||
| XM_017012845.1 | XP_016868334.1 | |||
| XM_017012846.1 | XP_016868335.1 | |||
| XM_017012850.1 | XP_016868339.1 | |||
| XM_017012851.1 | XP_016868340.1 | |||
| XM_017012852.1 | XP_016868341.1 | |||
| AB014605.2 | ||||
| AF038563.1 | AAC05370.1 | |||
| AK297353.1 | ||||
| BC150277.1 |
Target Gene Details
Entrez Gene ID: | 100874021 |
Gene Name: | MAGI2 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.7:79008988-79012277 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGI2-AS2 | NR_046689.1 | |||
| BX102296.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5803 | Chr.7:79008796 - 79040923 on Build GRCh38 | Insertion |
|
| nsv471642 | Chr.7:78919004 - 79088289 on Build GRCh38 | Loss |
|
| nsv831040 | Chr.7:78993879 - 79202682 on Build GRCh38 | Gain |
|
| nsv607659 | Chr.7:78863326 - 79854164 on Build GRCh38 | Gain |
|
| nsv521257 | Chr.7:79003173 - 79033109 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map