Assay Details
Target Gene Details
Entrez Gene ID: | 92140 |
Gene Name: | metadherin |
Gene Aliases: |
3D3, AEG-1, AEG1, LYRIC, LYRIC/3D3 |
Location: |
Chr.8:97643972-97730260 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTDH | NM_178812.3 | NP_848927.2 | ||
| XM_005251099.3 | XP_005251156.1 | |||
| XM_011517367.2 | XP_011515669.1 | |||
| XM_011517368.2 | XP_011515670.1 | |||
| XM_011517369.2 | XP_011515671.1 | |||
| XM_011517370.2 | XP_011515672.1 | |||
| XM_017013966.1 | XP_016869455.1 | |||
| XM_017013967.1 | XP_016869456.1 | |||
| XM_017013968.1 | XP_016869457.1 | |||
| AF411226.1 | AAO65771.1 | |||
| AF501310.1 | AAP30791.1 | |||
| AK000745.1 | ||||
| AY082966.1 | AAL92861.1 | |||
| AY974040.1 | AAX84832.1 | |||
| BC009324.2 | AAH09324.1 | |||
| BC014977.1 | ||||
| BC045642.1 | AAH45642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6325 | Chr.8:97619076 - 97663814 on Build GRCh38 | Deletion |
|
| esv2752271 | Chr.8:97536996 - 97728191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map