Assay Details
Target Gene Details
Entrez Gene ID: | 9228 |
Gene Name: | DLG associated protein 2 |
Gene Aliases: |
DAP2, SAPAP2 |
Location: |
Chr.8:1501366-1708476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP2 | NM_001277161.1 | NP_001264090.1 | ||
| NM_004745.4 | NP_004736.2 | |||
| AF119817.1 | AAF34344.1 | |||
| AF119818.1 | AAF34345.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469881 | Chr.8:1426471 - 1613783 on Build GRCh38 | Loss |
|
| nsv1024325 | Chr.8:651043 - 3203837 on Build GRCh38 | Gain |
|
| esv3576504 | Chr.8:951237 - 1906751 on Build GRCh38 | Gain |
|
| nsv1019956 | Chr.8:1474951 - 1514278 on Build GRCh38 | Gain |
|
| nsv609509 | Chr.8:1352944 - 1641722 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map