Assay Details
Target Gene Details
Entrez Gene ID: | 138046 |
Gene Name: | RALY RNA binding protein-like |
Gene Aliases: |
HNRPCL3 |
Location: |
Chr.8:84182629-84921844 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611646 | Chr.8:84136455 - 84200142 on Build GRCh38 | Loss |
|
| esv2752268 | Chr.8:83893110 - 84989630 on Build GRCh38 | Gain |
|
| nsv831374 | Chr.8:84191369 - 84396741 on Build GRCh38 | Loss |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117990142] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map