Assay Details
Target Gene Details
Entrez Gene ID: | 5157 |
Gene Name: | platelet derived growth factor receptor like |
Gene Aliases: |
PDGRL, PRLTS |
Location: |
Chr.8:17576433-17643133 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFRL | NM_006207.2 | NP_006198.1 | ||
| AK289450.1 | ||||
| BC010927.1 | AAH10927.1 | |||
| CR541952.1 | CAG46750.1 | |||
| D37965.1 | BAA07179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764066 | Chr.8:17530064 - 17677066 on Build GRCh38 | Gain+Loss |
|
| esv3616489 | Chr.8:17490020 - 17737549 on Build GRCh38 | Loss |
|
| nsv831246 | Chr.8:17430363 - 17621085 on Build GRCh38 | Gain |
|
| nsv831247 | Chr.8:17541589 - 17701918 on Build GRCh38 | Gain |
|
| esv3616492 | Chr.8:17598147 - 17735569 on Build GRCh38 | Loss |
|
| esv34867 | Chr.8:17488520 - 17725642 on Build GRCh38 | Loss |
|
| esv2759600 | Chr.8:17433671 - 17784021 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map