Assay Details
Target Gene Details
Entrez Gene ID: | 138046 |
Gene Name: | RALY RNA binding protein-like |
Gene Aliases: |
HNRPCL3 |
Location: |
Chr.8:84182629-84921844 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALYL | XM_017013063.1 | XP_016868552.1 | ||
| XM_017013065.1 | XP_016868554.1 | |||
| XM_017013071.1 | XP_016868560.1 | |||
| XM_017013074.1 | XP_016868563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611646 | Chr.8:84136455 - 84200142 on Build GRCh38 | Loss |
|
| esv2752268 | Chr.8:83893110 - 84989630 on Build GRCh38 | Gain |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map