Assay Details
Target Gene Details
Entrez Gene ID: | 138050 |
Gene Name: | heparan-alpha-glucosaminide N-acetyltransferase |
Gene Aliases: |
HGNAT, MPS3C, RP73, TMEM76 |
Location: |
Chr.8:43140449-43202827 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HGSNAT | NM_152419.2 | NP_689632.2 | ||
| XM_005273409.1 | XP_005273466.1 | |||
| XM_005273410.1 | XP_005273467.1 | |||
| XM_005273411.1 | XP_005273468.1 | |||
| XM_005273412.3 | XP_005273469.1 | |||
| XM_017013088.1 | XP_016868577.1 | |||
| XM_017013089.1 | XP_016868578.1 | |||
| AK304441.1 | ||||
| BC042037.1 | AAH42037.1 | |||
| DA395558.1 | ||||
| DB128369.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3617026 | Chr.8:43085930 - 43251904 on Build GRCh38 | Gain |
|
| dgv7187n100 | Chr.8:43081800 - 43968893 on Build GRCh38 | Gain |
|
| esv3891387 | Chr.8:42998571 - 43918067 on Build GRCh38 | Gain |
|
| nsv470208 | Chr.8:43082465 - 43936548 on Build GRCh38 | Gain |
|
| nsv831302 | Chr.8:43054340 - 43250377 on Build GRCh38 | Loss |
|
| nsv1032781 | Chr.8:42917251 - 43606341 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map