Assay Details
Target Gene Details
Entrez Gene ID: | 100129098 |
Gene Name: | translation initiation factor IF-2 |
Gene Aliases: |
- |
Location: |
Chr.8:54465594-54470426 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129098 | XM_017014124.1 | 2 | 860 | XP_016869613.1 |

Genomic Map