Assay Details
Target Gene Details
Entrez Gene ID: | 760 |
Gene Name: | carbonic anhydrase 2 |
Gene Aliases: |
CA-II, CAC, CAII, Car2, HEL-76, HEL-S-282 |
Location: |
Chr.8:85463902-85481492 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA2 | NM_000067.2 | NP_000058.1 | ||
| NM_001293675.1 | NP_001280604.1 | |||
| AK123309.1 | 1 | 381 | ||
| AK312978.1 | ||||
| BC011949.1 | AAH11949.1 | |||
| CD013933.1 | ||||
| CR536526.1 | CAG38763.1 | |||
| CR541875.1 | CAG46673.1 | |||
| FJ224289.1 | ||||
| GQ472235.1 | ||||
| J03037.1 | AAA51908.1 | |||
| M36532.1 | AAA51911.1 | |||
| Y00339.1 | CAA68426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831377 | Chr.8:85350878 - 85531912 on Build GRCh38 | Gain |
|
| nsv1028148 | Chr.8:85398341 - 85640901 on Build GRCh38 | Gain |
|
| esv2761206 | Chr.8:85398353 - 85556639 on Build GRCh38 | Gain |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map