Assay Details
Target Gene Details
| Entrez Gene ID: | 4956 | 
| Gene Name: | outer dense fiber of sperm tails 1 | 
| Gene Aliases: | CT133, HSPB10, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, RT7, SODF | 
| Location: | Chr.8:102551620-102561017 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ODF1 | NM_024410.3 | NP_077721.2 | ||
| AK315276.1 | ||||
| BC104456.1 | AAI04457.1 | |||
| BC104457.1 | AAI04458.1 | |||
| DB457574.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3618224 | Chr.8:102532605 - 102572593 on Build GRCh38 | Gain |  ODF1 | 
| esv2762761 | Chr.8:102480128 - 102613016 on Build GRCh38 | Gain |  ODF1 | 
| esv3576598 | Chr.8:102200959 - 102574604 on Build GRCh38 | Gain |  UBR5-AS1  ODF1  UBR5  RRM2B | 
| esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss |  LOC401472  ATP6V1C1  ODF1  AZIN1  LOC101927245  UBR5  KLF10  LOC107986873  AZIN1-AS1 | 
| nsv1024876 | Chr.8:102500557 - 102564430 on Build GRCh38 | Gain |  ODF1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top