Assay Details
Target Gene Details
Entrez Gene ID: | 4481 |
Gene Name: | macrophage scavenger receptor 1 |
Gene Aliases: |
CD204, SCARA1, SR-A, SRA, phSR1, phSR2 |
Location: |
Chr.8:16107878-16192791 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSR1 | NM_138715.2 | NP_619729.1 | ||
| NM_138716.2 | NP_619730.1 | |||
| AF037351.1 | AAC09251.1 | |||
| AK293217.1 | ||||
| BC063878.1 | AAH63878.1 | |||
| D90187.1 | BAA14208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831241 | Chr.8:16025262 - 16146736 on Build GRCh38 | Gain+Loss |
|
| esv3572569 | Chr.8:16110908 - 16114175 on Build GRCh38 | Loss |
|
| nsv1031894 | Chr.8:16084388 - 16157888 on Build GRCh38 | Gain |
|
| dgv1382e214 | Chr.8:16087881 - 16166383 on Build GRCh38 | Loss |
|
| esv3616432 | Chr.8:16089605 - 16164358 on Build GRCh38 | Gain |
|
| nsv1024623 | Chr.8:16017012 - 16146893 on Build GRCh38 | Gain |
|
| esv2764093 | Chr.8:16092468 - 16167255 on Build GRCh38 | Gain+Loss |
|
| esv3543130 | Chr.8:16087492 - 16166491 on Build GRCh38 | Deletion |
|
| dgv249e55 | Chr.8:16075727 - 16163959 on Build GRCh38 | Loss |
|
| nsv1023558 | Chr.8:16105571 - 16127519 on Build GRCh38 | Gain |
|
| dgv7109n100 | Chr.8:16086558 - 16167255 on Build GRCh38 | Loss |
|
| dgv12024n54 | Chr.8:16103682 - 16163959 on Build GRCh38 | Loss |
|
| dgv2050e212 | Chr.8:16086798 - 16166712 on Build GRCh38 | Loss |
|
| esv3891356 | Chr.8:16092262 - 16163959 on Build GRCh38 | Loss |
|
| esv3576529 | Chr.8:15792094 - 16396053 on Build GRCh38 | Gain |
|
| esv3616422 | Chr.8:16021420 - 16139525 on Build GRCh38 | Loss |
|
| nsv610576 | Chr.8:16070177 - 16381359 on Build GRCh38 | Gain |
|
| nsv1029399 | Chr.8:14978756 - 16296799 on Build GRCh38 | Gain |
|
| esv3616424 | Chr.8:16023868 - 16498645 on Build GRCh38 | Gain |
|
| esv2668117 | Chr.8:16087842 - 16166407 on Build GRCh38 | Deletion |
|
| esv3572572 | Chr.8:16059385 - 16136348 on Build GRCh38 | Loss |
|
| dgv12022n54 | Chr.8:16085204 - 16171983 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145329024,rs78105795] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map