Assay Details
Target Gene Details
Entrez Gene ID: | 28 |
Gene Name: | ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase) |
Gene Aliases: |
A3GALNT, A3GALT1, GTB, NAGAT |
Location: |
Chr.9:133255176-133275214 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABO | NM_020469.2 | NP_065202.2 | ||
| AF134412.1 | AAD26572.1 | |||
| AF134413.1 | AAD26573.1 | |||
| AF134414.1 | AAD26574.1 | |||
| AF134415.1 | ||||
| AF134416.1 | ||||
| AK302203.1 | ||||
| AV761252.1 | ||||
| BC069595.1 | AAH69595.1 | |||
| BC069814.1 | AAH69814.1 | |||
| BC111575.1 | ||||
| DQ139865.1 | ||||
| FR691751.1 | ||||
| FR691752.1 | ||||
| J05175.1 | AAA36792.1 | |||
| JN791441.1 | ||||
| JX398330.1 | ||||
| JX398331.1 | ||||
| JX398332.1 | ||||
| JX398333.1 | ||||
| JX519569.1 | ||||
| JX519570.1 | ||||
| KT282967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv469905 | Chr.9:133174393 - 133290774 on Build GRCh38 | Gain |
|
| dgv951n27 | Chr.9:133156364 - 133320327 on Build GRCh38 | Gain |
|
| nsv615616 | Chr.9:133254625 - 133259604 on Build GRCh38 | Loss |
|
| nsv951798 | Chr.9:133254614 - 133263597 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map