Assay Details
Target Gene Details
Entrez Gene ID: | 57582 |
Gene Name: | potassium sodium-activated channel subfamily T member 1 |
Gene Aliases: |
EIEE14, ENFL5, KCa4.1, SLACK, Slo2.2, bA100C15.2 |
Location: |
Chr.9:135700807-135793147 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNT1 | NM_001272003.1 | NP_001258932.1 | ||
| NM_020822.2 | NP_065873.2 | |||
| XM_011518877.2 | XP_011517179.1 | |||
| XM_011518878.2 | XP_011517180.1 | |||
| XM_011518879.2 | XP_011517181.1 | |||
| AB037843.1 | ||||
| AK127272.1 | ||||
| BC136618.1 | ||||
| BC171770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831756 | Chr.9:135648434 - 135835577 on Build GRCh38 | Loss |
|
| nsv469915 | Chr.9:135699384 - 135739808 on Build GRCh38 | Gain |
|
| dgv12991n54 | Chr.9:135689001 - 135752357 on Build GRCh38 | Loss |
|
| nsv951201 | Chr.9:135647755 - 135770054 on Build GRCh38 | Deletion |
|
| nsv825171 | Chr.9:135693613 - 135767026 on Build GRCh38 | Gain |
|
| nsv615927 | Chr.9:135714078 - 135738597 on Build GRCh38 | Loss |
|
| nsv517335 | Chr.9:135655608 - 135803389 on Build GRCh38 | Loss |
|
| dgv12994n54 | Chr.9:135711894 - 135789055 on Build GRCh38 | Gain |
|
| nsv469914 | Chr.9:135667037 - 135789055 on Build GRCh38 | Loss |
|
| dgv954n27 | Chr.9:135698139 - 135761806 on Build GRCh38 | Loss |
|
| dgv12992n54 | Chr.9:135689825 - 135725457 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map