Assay Details
Target Gene Details
Entrez Gene ID: | 10422 |
Gene Name: | UBA domain containing 1 |
Gene Aliases: |
GBDR1, UBADC1 |
Location: |
Chr.9:135932969-135961380 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBAC1 | NM_016172.2 | NP_057256.2 | ||
| AF038200.1 | ||||
| AF068195.1 | AAC21559.1 | |||
| AF176796.1 | AAD51084.1 | |||
| BC004967.1 | AAH04967.1 | |||
| BC011822.1 | AAH11822.1 | |||
| BX647463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12995n54 | Chr.9:135903671 - 135992854 on Build GRCh38 | Loss |
|
| nsv831757 | Chr.9:135885814 - 136184089 on Build GRCh38 | Loss |
|
| dgv7728n100 | Chr.9:135864526 - 135949196 on Build GRCh38 | Gain |
|
| nsv825173 | Chr.9:135892081 - 136495763 on Build GRCh38 | Gain |
|
| nsv615928 | Chr.9:135885466 - 136002028 on Build GRCh38 | Gain |
|
| nsv951202 | Chr.9:135770355 - 136558248 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map