Assay Details
Target Gene Details
Entrez Gene ID: | 64077 |
Gene Name: | phospholysine phosphohistidine inorganic pyrophosphate phosphatase |
Gene Aliases: |
HDHD2B |
Location: |
Chr.10:124461772-124614141 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHPP | NM_001167880.1 | NP_001161352.1 | ||
| NM_001318331.1 | NP_001305260.1 | |||
| NM_001318332.1 | NP_001305261.1 | |||
| NM_022126.3 | NP_071409.3 | |||
| XM_005270026.3 | XP_005270083.1 | |||
| XM_006717943.1 | XP_006718006.1 | |||
| XM_011540058.2 | XP_011538360.1 | |||
| XM_017016509.1 | XP_016871998.1 | |||
| XM_017016510.1 | XP_016871999.1 | |||
| XM_017016511.1 | XP_016872000.1 | |||
| AK055532.1 | ||||
| AW631352.1 | ||||
| BC110344.1 | AAI10345.1 | |||
| BC113629.1 | ||||
| BC113631.1 | ||||
| BM541880.1 | ||||
| HY154569.1 |
More Information
Set Membership: |
|

Genomic Map