Assay Details
Target Gene Details
Entrez Gene ID: | 317662 |
Gene Name: | family with sequence similarity 149 member B1 |
Gene Aliases: |
KIAA0974 |
Location: |
Chr.10:73168090-73242181 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM149B1 | NM_173348.1 | NP_775483.1 | ||
| XM_005269744.2 | XP_005269801.1 | |||
| XM_005269745.2 | XP_005269802.1 | |||
| XM_005269747.3 | XP_005269804.1 | |||
| XM_011539737.2 | XP_011538039.1 | |||
| XM_011539740.2 | XP_011538042.1 | |||
| XM_017016164.1 | XP_016871653.1 | |||
| XM_017016165.1 | XP_016871654.1 | |||
| XM_017016166.1 | XP_016871655.1 | |||
| XM_017016167.1 | XP_016871656.1 | |||
| AB023191.1 | BAA76818.1 | |||
| AK303436.1 | ||||
| BC015394.1 | AAH15394.1 | |||
| DC340839.1 |

Genomic Map