Assay Details
Target Gene Details
Entrez Gene ID: | 57705 |
Gene Name: | WDFY family member 4 |
Gene Aliases: |
C10orf64 |
Location: |
Chr.10:48684815-48982956 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY4 | NM_020945.1 | NP_065996.1 | ||
| XM_005270004.3 | XP_005270061.1 | |||
| XM_011539986.2 | XP_011538288.2 | |||
| XM_011539987.2 | XP_011538289.2 | |||
| XM_011539988.2 | XP_011538290.1 | |||
| XM_011539990.2 | XP_011538292.1 | |||
| XM_011539991.2 | XP_011538293.1 | |||
| XM_011539992.2 | XP_011538294.1 | |||
| XM_017016463.1 | XP_016871952.1 | |||
| XM_017016465.1 | XP_016871954.1 | |||
| AK127650.1 | BAC87073.1 | |||
| BC034937.1 | AAH34937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142937944] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map