Assay Details
Target Gene Details
Entrez Gene ID: | 1326 |
Gene Name: | mitogen-activated protein kinase kinase kinase 8 |
Gene Aliases: |
AURA2, COT, EST, ESTF, MEKK8, TPL2, Tpl-2, c-COT |
Location: |
Chr.10:30434021-30461833 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K8 | NM_001244134.1 | NP_001231063.1 | ||
| NM_001320961.1 | NP_001307890.1 | |||
| NM_005204.3 | NP_005195.2 | |||
| XM_017015708.1 | XP_016871197.1 | |||
| XM_017015709.1 | 1 | 1125 | XP_016871198.1 | |
| XM_017015712.1 | XP_016871201.1 | |||
| XM_017015713.1 | XP_016871202.1 | |||
| XM_017015714.1 | XP_016871203.1 | |||
| AK290320.1 | ||||
| AK313295.1 | ||||
| AL547407.3 | ||||
| BI753824.1 | ||||
| BP352229.1 | ||||
| D14497.1 | BAA03387.1 | |||
| DA308072.1 | ||||
| DB271664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161936 | Chr.10:30113251 - 31078102 on Build GRCh38 | Duplication |
|
| nsv831821 | Chr.10:30346221 - 30536200 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map