Assay Details
Target Gene Details
Entrez Gene ID: | 100130887 |
Gene Name: | ATE1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121928312-121951965 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATE1-AS1 | NR_120495.1 | |||
AI634505.1 | ||||
BF434328.1 |
