Assay Details
Target Gene Details
Entrez Gene ID: | 64376 |
Gene Name: | IKAROS family zinc finger 5 |
Gene Aliases: |
PEGASUS, ZNFN1A5 |
Location: |
Chr.10:122990806-123008850 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IKZF5 | NM_001271840.1 | NP_001258769.1 | ||
NR_073475.1 | ||||
NR_073476.1 | ||||
NR_073477.1 | ||||
NR_073478.1 | ||||
NR_073479.1 | ||||
XM_006717947.3 | XP_006718010.1 | |||
XM_017016550.1 | XP_016872039.1 | |||
XM_017016551.1 | XP_016872040.1 | |||
XM_017016552.1 | XP_016872041.1 | |||
XM_017016553.1 | XP_016872042.1 | |||
XM_017016554.1 | XP_016872043.1 | |||
XM_017016555.1 | XP_016872044.1 | |||
AK026626.1 | BAB15512.1 | |||
AK122899.1 | ||||
AK226143.1 | ||||
BX647958.1 | CAH56199.1 | |||
CR749800.1 | CAH18660.1 | |||
DA218215.1 | ||||
DA227834.1 | ||||
DB080324.1 | ||||
DB224006.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv7586 | Chr.10:122982687 - 123016052 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
