Assay Details
Target Gene Details
Entrez Gene ID: | 8559 |
Gene Name: | pre-mRNA processing factor 18 |
Gene Aliases: |
PRP18, hPrp18 |
Location: |
Chr.10:13586811-13645279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF18 | NM_003675.3 | NP_003666.1 | ||
| XM_011519754.2 | XP_011518056.1 | |||
| XM_017016860.1 | XP_016872349.1 | |||
| AK314814.1 | ||||
| AU142253.1 | ||||
| BC000794.1 | AAH00794.1 | |||
| U51990.1 | AAB41490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831789 | Chr.10:13536081 - 13716895 on Build GRCh38 | Gain+Loss |
|
| esv3891761 | Chr.10:13544627 - 13695972 on Build GRCh38 | Gain |
|
| esv2761575 | Chr.10:13581531 - 13659502 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map