Assay Details
Target Gene Details
Entrez Gene ID: | 100126784 |
Gene Name: | uncharacterized LOC100126784 |
Gene Aliases: |
- |
Location: |
Chr.11:19710934-19714672 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100126784 | NR_015384.2 | 1 | 1658 | |
| DA492854.1 | 1 | 181 |
Target Gene Details
Entrez Gene ID: | 89797 |
Gene Name: | neuron navigator 2 |
Gene Aliases: |
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2 |
Location: |
Chr.11:19345200-20121601 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAV2 | NM_001111018.1 | NP_001104488.1 | ||
| XM_006718364.3 | XP_006718427.1 | |||
| XM_006718365.3 | XP_006718428.1 | |||
| XM_011520444.2 | XP_011518746.1 | |||
| XM_011520445.2 | XP_011518747.1 | |||
| XM_011520446.2 | XP_011518748.1 | |||
| XM_011520447.2 | XP_011518749.1 | |||
| XM_011520448.2 | XP_011518750.1 | |||
| XM_011520449.2 | XP_011518751.1 | |||
| XM_011520450.2 | XP_011518752.1 | |||
| XM_011520452.2 | XP_011518754.1 | |||
| XM_017018520.1 | XP_016874009.1 | |||
| XM_017018521.1 | XP_016874010.1 | |||
| XM_017018522.1 | XP_016874011.1 | |||
| XM_017018524.1 | XP_016874013.1 | |||
| AB063115.1 | BAC00853.1 | |||
| BX459843.2 | 1 | 1010 |
More Information
Set Membership: |
|

Genomic Map