Assay Details
Target Gene Details
Entrez Gene ID: | 120114 |
Gene Name: | FAT atypical cadherin 3 |
Gene Aliases: |
CDHF15, CDHR10, hFat3 |
Location: |
Chr.11:92224636-92896473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAT3 | XM_017017178.1 | XP_016872667.1 | ||
| XM_017017179.1 | XP_016872668.1 | |||
| XM_017017180.1 | XP_016872669.1 | |||
| XM_017017181.1 | XP_016872670.1 | |||
| XM_017017182.1 | XP_016872671.1 | |||
| XM_017017183.1 | XP_016872672.1 | |||
| XM_017017184.1 | XP_016872673.1 | |||
| XM_017017186.1 | XP_016872675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555902 | Chr.11:92195159 - 92251289 on Build GRCh38 | Loss |
|
| nsv832229 | Chr.11:92151270 - 92341658 on Build GRCh38 | Gain |
|
| esv3580015 | Chr.11:91820356 - 92847634 on Build GRCh38 | Gain |
|
| nsv832230 | Chr.11:92219476 - 92379180 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map