Assay Details
Target Gene Details
Entrez Gene ID: | 26011 |
Gene Name: | teneurin transmembrane protein 4 |
Gene Aliases: |
Doc4, ETM5, ODZ4, TNM4, Ten-M4 |
Location: |
Chr.11:78653283-79441019 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TENM4 | NM_001098816.2 | NP_001092286.2 | ||
| XM_011544933.2 | XP_011543235.1 | |||
| XM_017017525.1 | XP_016873014.1 | |||
| XM_017017526.1 | XP_016873015.1 | |||
| AB037723.3 | BAA92540.3 | |||
| AK127101.1 | ||||
| AK127705.1 | ||||
| BX537983.1 | CAD97943.1 | |||
| BX640737.1 | CAE45850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483084 | Chr.11:78645595 - 78818126 on Build GRCh38 | Loss |
|
| esv34127 | Chr.11:78661992 - 79102008 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map