Assay Details
Target Gene Details
Entrez Gene ID: | 79832 |
Gene Name: | glutamine and serine rich 1 |
Gene Aliases: |
- |
Location: |
Chr.11:32892826-32980270 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QSER1 | NM_001076786.2 | NP_001070254.1 | ||
| XM_006718325.3 | XP_006718388.3 | |||
| XM_017018327.1 | XP_016873816.1 | |||
| XM_017018328.1 | XP_016873817.1 | |||
| XM_017018329.1 | XP_016873818.1 | |||
| XM_017018330.1 | XP_016873819.1 | |||
| AK126023.1 | BAC86397.1 | |||
| BC112935.1 | AAI12936.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040484 | Chr.11:32929520 - 33071658 on Build GRCh38 | Gain |
|
| esv2759815 | Chr.11:32693097 - 33028394 on Build GRCh38 | Gain |
|
| esv34742 | Chr.11:32774546 - 32982478 on Build GRCh38 | Gain |
|

Genomic Map