Assay Details
Target Gene Details
Entrez Gene ID: | 22997 |
Gene Name: | immunoglobulin superfamily member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:133908564-133956960 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF9B | NM_001277285.1 | 18 | 3366 | NP_001264214.1 |
| XM_006718795.3 | 7 | 1880 | XP_006718858.1 | |
| XM_011542691.2 | 19 | 4053 | XP_011540993.1 | |
| XM_011542692.2 | 19 | 4041 | XP_011540994.1 | |
| XM_011542694.2 | 19 | 4056 | XP_011540996.1 | |
| XM_011542695.2 | 18 | 3584 | XP_011540997.1 | |
| XM_017017393.1 | 18 | 4071 | XP_016872882.1 | |
| XM_017017394.1 | 19 | 4056 | XP_016872883.1 | |
| XM_017017395.1 | 19 | 4055 | XP_016872884.1 | |
| XM_017017396.1 | 12 | 3395 | XP_016872885.1 | |
| AB028953.1 | 6 | 1608 | BAA82982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051677 | Chr.11:133862255 - 133955026 on Build GRCh38 | Gain |
|
| nsv952765 | Chr.11:133891206 - 133952805 on Build GRCh38 | Deletion |
|
| nsv826144 | Chr.11:133920301 - 134020350 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map