Assay Details
Target Gene Details
Entrez Gene ID: | 219988 |
Gene Name: | PAT1 homolog 1, processing body mRNA decay factor |
Gene Aliases: |
Pat1b, hPat1b |
Location: |
Chr.11:59636716-59669040 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PATL1 | NM_152716.2 | NP_689929.2 | ||
| XM_005273833.4 | XP_005273890.1 | |||
| XM_005273834.1 | XP_005273891.1 | |||
| XM_011544811.2 | XP_011543113.1 | |||
| XM_017017339.1 | XP_016872828.1 | |||
| AK094193.1 | BAC04305.1 | |||
| AK127649.1 | ||||
| AK127943.1 | ||||
| AL831992.2 | CAD89916.1 | |||
| BC065264.1 | ||||
| BC109038.1 | AAI09039.1 | |||
| BC109039.1 | AAI09040.1 | |||
| BC111047.1 | AAI11048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832172 | Chr.11:59628934 - 59663642 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map