Assay Details
Target Gene Details
Entrez Gene ID: | 10235 |
Gene Name: | RAS guanyl releasing protein 2 |
Gene Aliases: |
CALDAG-GEFI, CDC25L |
Location: |
Chr.11:64726911-64745456 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASGRP2 | NM_001098670.1 | NP_001092140.1 | ||
| NM_001098671.1 | NP_001092141.1 | |||
| NM_001318398.1 | NP_001305327.1 | |||
| NM_153819.1 | NP_722541.1 | |||
| XM_005273707.3 | XP_005273764.3 | |||
| XM_011544718.1 | XP_011543020.1 | |||
| XM_011544720.1 | XP_011543022.1 | |||
| XM_011544721.1 | XP_011543023.1 | |||
| XM_011544722.1 | XP_011543024.1 | |||
| XM_011544723.2 | XP_011543025.1 | |||
| XM_017017082.1 | 1 | 1064 | XP_016872571.1 | |
| XM_017017083.1 | 1 | 1064 | XP_016872572.1 | |
| XM_017017084.1 | XP_016872573.1 | |||
| AF043722.1 | AAF07219.1 | |||
| AF043723.1 | AAF07220.1 | |||
| AF081194.1 | AAC79698.1 | |||
| AK092882.1 | 1 | 679 | ||
| AK122694.1 | ||||
| AK124023.1 | 1 | 1479 | ||
| BC110306.1 | AAI10307.1 | |||
| BC117151.1 | ||||
| DA286186.1 | ||||
| DA529151.1 | ||||
| DB498812.1 | ||||
| U78170.1 | AAD12741.1 | |||
| Y12336.1 | CAA73005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528506 | Chr.11:64577976 - 64933558 on Build GRCh38 | Loss |
|
| dgv1965n54 | Chr.11:64566642 - 64806117 on Build GRCh38 | Loss |
|
| dgv153n27 | Chr.11:64709578 - 64817487 on Build GRCh38 | Loss |
|
| nsv832189 | Chr.11:64719711 - 64852727 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map