Assay Details
Target Gene Details
Entrez Gene ID: | 9866 |
Gene Name: | tripartite motif containing 66 |
Gene Aliases: |
C11orf29, TIF1D, TIF1DELTA |
Location: |
Chr.11:8612037-8683217 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM66 | NM_014818.1 | 20 | 7116 | NP_055633.1 |
| XM_006718397.2 | 25 | 8054 | XP_006718460.1 | |
| XM_006718398.3 | 21 | 8666 | XP_006718461.1 | |
| XM_011520504.1 | 26 | 8097 | XP_011518806.1 | |
| XM_011520507.2 | 25 | 8018 | XP_011518809.1 | |
| XM_011520508.1 | 25 | 8097 | XP_011518810.1 | |
| XM_011520509.1 | 24 | 7979 | XP_011518811.1 | |
| XM_011520510.1 | 25 | 7970 | XP_011518812.1 | |
| XM_011520511.1 | 25 | 7943 | XP_011518813.1 | |
| XM_011520512.1 | 21 | 7318 | XP_011518814.1 | |
| XM_011520513.1 | 21 | 7316 | XP_011518815.1 | |
| XM_011520514.2 | 21 | 7272 | XP_011518816.1 | |
| XM_011520515.1 | 22 | 7485 | XP_011518817.1 | |
| XM_011520516.1 | 22 | 7485 | XP_011518818.1 | |
| XM_011520517.2 | 22 | 8807 | XP_011518819.1 | |
| XM_011520518.1 | 22 | 7455 | XP_011518820.1 | |
| XM_011520519.1 | 21 | 7343 | XP_011518821.1 | |
| XM_011520522.2 | 20 | 8521 | XP_011518824.1 | |
| XM_011520523.2 | 21 | 9403 | XP_011518825.1 | |
| XM_011520526.2 | 13 | 6372 | XP_011518828.1 | |
| XM_011520527.1 | 13 | 5640 | XP_011518829.1 | |
| XM_017018629.1 | 25 | 8447 | XP_016874118.1 | |
| XM_017018630.1 | 20 | 7409 | XP_016874119.1 | |
| XM_017018631.1 | 20 | 7561 | XP_016874120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751035 | Chr.11:8410107 - 8663904 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map