Assay Details
Target Gene Details
Entrez Gene ID: | 387804 |
Gene Name: | V-set and transmembrane domain containing 5 |
Gene Aliases: |
C11orf90 |
Location: |
Chr.11:93817315-93850653 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM5 | NM_001144871.1 | NP_001138343.1 | ||
| DY654877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892068 | Chr.11:93819689 - 93897325 on Build GRCh38 | Gain |
|
| esv3627294 | Chr.11:93485306 - 93944317 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115169147] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map