Assay Details
Target Gene Details
Entrez Gene ID: | 143872 |
Gene Name: | Rho GTPase activating protein 42 |
Gene Aliases: |
GRAF3 |
Location: |
Chr.11:100687676-100993941 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP42 | NM_152432.2 | NP_689645.2 | ||
| AI672473.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1265n100 | Chr.11:100017658 - 100758405 on Build GRCh38 | Gain |
|
| nsv832246 | Chr.11:100555994 - 100736226 on Build GRCh38 | Gain |
|
| nsv520995 | Chr.11:100293225 - 100810480 on Build GRCh38 | Gain |
|
| nsv556122 | Chr.11:100022550 - 100715158 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map