Assay Details
Target Gene Details
Entrez Gene ID: | 9415 |
Gene Name: | fatty acid desaturase 2 |
Gene Aliases: |
D6D, DES6, FADSD6, LLCDL2, SLL0262, TU13 |
Location: |
Chr.11:61816203-61867354 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADS2 | NM_001281501.1 | NP_001268430.1 | ||
| NM_001281502.1 | NP_001268431.1 | |||
| AF108658.1 | AAG43192.1 | |||
| AK074991.1 | ||||
| AK290291.1 | ||||
| AK299762.1 | ||||
| HY090295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map