Assay Details
Target Gene Details
Entrez Gene ID: | 25977 |
Gene Name: | NECAP endocytosis associated 1 |
Gene Aliases: |
EIEE21 |
Location: |
Chr.12:8082211-8097777 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NECAP1 | NM_015509.3 | NP_056324.2 | ||
NR_024260.1 | ||||
AK074858.1 | BAC11250.1 | |||
AK074880.1 | BAC11264.1 | |||
AK074923.1 | BAC11296.1 | |||
AK075013.1 | BAC11352.1 | |||
AK290537.1 | ||||
AK303675.1 | ||||
AK314424.1 | ||||
AL050272.1 | CAB43373.2 | |||
BC002888.2 | AAH02888.1 | |||
BC110876.1 | AAI10877.1 | |||
DA496321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759877 | Chr.12:7704223 - 8471391 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1045521 | Chr.12:7970629 - 8090250 on Build GRCh38 | Gain |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
