Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C12orf60 |
Target Gene Details
Entrez Gene ID: | 101928290 |
Gene Name: | PLBD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:14567732-14619755 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLBD1-AS1 | NR_120465.1 | |||
AI922058.1 | ||||
CB048614.1 | ||||
DB284711.1 |
