Assay Details
Target Gene Details
Entrez Gene ID: | 64786 |
Gene Name: | TBC1 domain family member 15 |
Gene Aliases: |
RAB7-GAP |
Location: |
Chr.12:71839707-71926849 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759907 | Chr.12:71708684 - 71914393 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv559329 | Chr.12:71748682 - 71886804 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3892218 | Chr.12:71785292 - 71892336 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3629916 | Chr.12:71783025 - 71889798 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv818924 | Chr.12:71788125 - 71886804 on Build GRCh38 | Gain |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
