Assay Details
Target Gene Details
Entrez Gene ID: | 7421 |
Gene Name: | vitamin D (1,25- dihydroxyvitamin D3) receptor |
Gene Aliases: |
NR1I1, PPP1R163 |
Location: |
Chr.12:47841537-47905031 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VDR | NM_000376.2 | NP_000367.1 | ||
| NM_001017535.1 | NP_001017535.1 | |||
| NM_001017536.1 | NP_001017536.1 | |||
| XM_006719587.3 | XP_006719650.1 | |||
| XM_011538720.2 | XP_011537022.1 | |||
| AB307700.1 | ||||
| AF026260.1 | AAB95155.1 | |||
| AK225844.1 | ||||
| AK299446.1 | ||||
| AK312267.1 | ||||
| BC060832.1 | AAH60832.1 | |||
| BX355584.2 | ||||
| CF138099.1 | ||||
| HQ692836.1 | ||||
| J03258.1 | AAA61273.1 | |||
| X67482.1 | CAA47824.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116299778,rs117913411] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map