Assay Details
Target Gene Details
Entrez Gene ID: | 57701 |
Gene Name: | NCK associated protein 5 like |
Gene Aliases: |
Cep169, FP1193, KIAA1602 |
Location: |
Chr.12:49791139-49828750 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCKAP5L | NM_001037806.3 | 8 | 1874 | NP_001032895.2 |
| XM_005269050.3 | 8 | 2199 | XP_005269107.1 | |
| XM_006719525.2 | 8 | 2169 | XP_006719588.1 | |
| XM_011538610.2 | 9 | 1953 | XP_011536912.1 | |
| XM_011538611.2 | 9 | 1923 | XP_011536913.1 | |
| AB046822.1 | 1 | 815 | BAB13428.1 | |
| BC033253.1 | 1 | 390 | AAH33253.1 |
More Information
Set Membership: |
|

Genomic Map