Assay Details
Target Gene Details
Entrez Gene ID: | 54509 |
Gene Name: | ras homolog family member F, filopodia associated |
Gene Aliases: |
ARHF, RIF |
Location: |
Chr.12:121777754-121793688 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOF | NM_019034.2 | NP_061907.2 | ||
| AF239923.1 | AAG24952.1 | |||
| AK000254.1 | BAA91034.1 | |||
| AK289482.1 | ||||
| BC018208.1 | AAH18208.1 | |||
| BG574506.1 | ||||
| BQ053707.1 | ||||
| BU527972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160065 | Chr.12:121780583 - 121859371 on Build GRCh38 | Deletion |
|
| esv3892268 | Chr.12:121747658 - 121845875 on Build GRCh38 | Loss |
|
| esv2746493 | Chr.12:121791713 - 121792629 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map