Assay Details
Target Gene Details
Entrez Gene ID: | 55846 |
Gene Name: | integrin alpha FG-GAP repeat containing 2 |
Gene Aliases: |
MDS028 |
Location: |
Chr.12:2812621-2832217 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITFG2 | NM_018463.3 | NP_060933.3 | ||
| NR_130744.1 | ||||
| XM_005253707.4 | XP_005253764.1 | |||
| XM_005253709.4 | XP_005253766.1 | |||
| XM_017019641.1 | XP_016875130.1 | |||
| AK027483.1 | BAB55145.1 | |||
| AK130319.1 | ||||
| AK291110.1 | ||||
| AK297189.1 | ||||
| AK300905.1 | ||||
| AY599883.1 | AAT09136.1 | |||
| BC006552.2 | AAH06552.1 | |||
| BC013399.2 | AAH13399.1 | |||
| BI261823.1 | ||||
| KU178719.1 | ||||
| KU178720.1 | ||||
| KU178721.1 | ||||
| KU178722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628322 | Chr.12:2796908 - 2837171 on Build GRCh38 | Gain |
|
| esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain |
|
| nsv826194 | Chr.12:2777359 - 2817185 on Build GRCh38 | Gain |
|
| dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss |
|
| nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map