Assay Details
Target Gene Details
Entrez Gene ID: | 121512 |
Gene Name: | FYVE, RhoGEF and PH domain containing 4 |
Gene Aliases: |
CMT4H, FRABP, ZFYVE6 |
Location: |
Chr.12:32399523-32646050 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD4 | XM_005253304.4 | XP_005253361.1 | ||
| XM_005253309.1 | XP_005253366.1 | |||
| XM_017018803.1 | XP_016874292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469347 | Chr.12:32402912 - 32449145 on Build GRCh38 | Gain |
|
| esv2751096 | Chr.12:32127406 - 32549850 on Build GRCh38 | Gain |
|
| esv3580220 | Chr.12:32404645 - 32462728 on Build GRCh38 | Loss |
|
| dgv265e214 | Chr.12:32419473 - 32453826 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map