Assay Details
Target Gene Details
Entrez Gene ID: | 6540 |
Gene Name: | solute carrier family 6 member 13 |
Gene Aliases: |
GAT-2, GAT2, GAT3 |
Location: |
Chr.12:220621-262930 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A13 | NM_001190997.2 | NP_001177926.1 | ||
| NM_016615.4 | NP_057699.2 | |||
| XM_006719008.3 | XP_006719071.1 | |||
| XM_011521012.2 | XP_011519314.1 | |||
| XM_011521013.1 | XP_011519315.1 | |||
| XM_011521014.1 | XP_011519316.1 | |||
| XM_017019842.1 | XP_016875331.1 | |||
| XM_017019843.1 | XP_016875332.1 | |||
| XM_017019844.1 | XP_016875333.1 | |||
| XM_017019845.1 | XP_016875334.1 | |||
| AK296127.1 | ||||
| AK296209.1 | ||||
| AK313511.1 | ||||
| BC022392.1 | AAH22392.1 | |||
| U76343.2 | AAF64247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556855 | Chr.12:154673 - 226756 on Build GRCh38 | Loss |
|
| nsv556861 | Chr.12:184347 - 251448 on Build GRCh38 | Loss |
|
| nsv832302 | Chr.12:57392 - 234835 on Build GRCh38 | Gain |
|
| nsv1050781 | Chr.12:212187 - 674733 on Build GRCh38 | Loss |
|
| nsv556857 | Chr.12:170292 - 268738 on Build GRCh38 | Gain |
|
| esv3628226 | Chr.12:157297 - 251505 on Build GRCh38 | Gain |
|
| esv3628221 | Chr.12:114421 - 232803 on Build GRCh38 | Gain |
|
| dgv1330n100 | Chr.12:45001 - 233978 on Build GRCh38 | Gain |
|
| nsv470253 | Chr.12:218255 - 250297 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73038087] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map