Assay Details
Target Gene Details
| Entrez Gene ID: | 100507091 | 
| Gene Name: | uncharacterized LOC100507091 | 
| Gene Aliases: | - | 
| Location: | Chr.12:123081384-123084744 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LOC100507091 | NR_038290.1 | 3 | 1270 | |
| AK093493.1 | 3 | 1270 | 
Target Gene Details
| Entrez Gene ID: | 57605 | 
| Gene Name: | phosphatidylinositol transfer protein membrane associated 2 | 
| Gene Aliases: | NIR-3, NIR3, RDGB2, RDGBA2 | 
| Location: | Chr.12:122983480-123110489 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| PITPNM2 | NM_001300801.1 | NP_001287730.1 | ||
| NM_020845.2 | NP_065896.1 | |||
| AB040890.2 | ||||
| BC141813.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |  ABCB9  PITPNM2  OGFOD2  MIR4304  LOC100507091  ARL6IP4 | 
| nsv560492 | Chr.12:122943026 - 123090291 on Build GRCh38 | Loss |  ABCB9  PITPNM2  OGFOD2  MIR4304  LOC100507091  ARL6IP4 | 
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |  HIP1R  ABCB9  PITPNM2  VPS37B  OGFOD2  MPHOSPH9  MIR4304  LOC100507091  ARL6IP4 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top