Assay Details
Target Gene Details
Entrez Gene ID: | 144404 |
Gene Name: | transmembrane protein 120B |
Gene Aliases: |
- |
Location: |
Chr.12:121712752-121782070 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM120B | NM_001080825.2 | NP_001074294.2 | ||
| XM_017018853.1 | XP_016874342.1 | |||
| XM_017018854.1 | XP_016874343.1 | |||
| AK126568.1 | ||||
| BC127768.1 | ||||
| BC127769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892268 | Chr.12:121747658 - 121845875 on Build GRCh38 | Loss |
|
| dgv882n106 | Chr.12:121750394 - 121751794 on Build GRCh38 | Deletion |
|
| nsv560489 | Chr.12:121747710 - 121752479 on Build GRCh38 | Loss |
|
| nsv560472 | Chr.12:121747352 - 121752479 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map