Assay Details
Target Gene Details
Entrez Gene ID: | 6144 |
Gene Name: | ribosomal protein L21 |
Gene Aliases: |
HYPT12, L21 |
Location: |
Chr.13:27251555-27256568 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL21 |

Genomic Map