Assay Details
Target Gene Details
Entrez Gene ID: | 6144 |
Gene Name: | ribosomal protein L21 |
Gene Aliases: |
HYPT12, L21 |
Location: |
Chr.13:27251555-27256568 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976098 | Chr.13:27253748 - 27256597 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111462631] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map