Assay Details
Target Gene Details
Entrez Gene ID: | 10166 |
Gene Name: | solute carrier family 25 member 15 |
Gene Aliases: |
D13S327, HHH, ORC1, ORNT1 |
Location: |
Chr.13:40789411-40812460 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A15 | NM_014252.3 | NP_055067.1 | ||
| AF112968.1 | AAD45238.1 | |||
| AF177333.1 | AAG17977.1 | |||
| AK296862.1 | ||||
| AK313602.1 | ||||
| AL538057.3 | ||||
| BC002702.2 | AAH02702.1 | |||
| BI461478.1 | ||||
| DB505424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892332 | Chr.13:40619338 - 40820761 on Build GRCh38 | Gain |
|
| nsv974081 | Chr.13:40791932 - 40795053 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map