Assay Details
Target Gene Details
Entrez Gene ID: | 10810 |
Gene Name: | WAS protein family member 3 |
Gene Aliases: |
Brush-1, SCAR3, WAVE3 |
Location: |
Chr.13:26557703-26688945 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WASF3 | NM_001291965.1 | NP_001278894.1 | ||
| NM_006646.5 | NP_006637.2 | |||
| AF454702.1 | AAL51032.1 | |||
| AK289845.1 | ||||
| BC050283.1 | AAH50283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826597 | Chr.13:26552349 - 26560300 on Build GRCh38 | Gain |
|
| esv2759928 | Chr.13:26426455 - 26593241 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75695435] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map