Assay Details
Target Gene Details
Entrez Gene ID: | 729420 |
Gene Name: | LMO7 downstream neighbor |
Gene Aliases: |
C13orf45 |
Location: |
Chr.13:75869019-75883888 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO7DN | NM_001257995.1 | NP_001244924.1 | ||
| XM_011535208.2 | XP_011533510.1 | |||
| AK092698.1 | BAC03949.1 |
Target Gene Details
Entrez Gene ID: | 104326189 |
Gene Name: | LMO7DN intronic transcript 1 |
Gene Aliases: |
C13orf45-IT1, TCONS_00021569 |
Location: |
Chr.13:75876886-75881127 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO7DN-IT1 | NR_126373.1 | |||
| AI825938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832649 | Chr.13:75850736 - 76083755 on Build GRCh38 | Loss |
|
| nsv832646 | Chr.13:75726292 - 75908966 on Build GRCh38 | Gain |
|
| esv2759950 | Chr.13:75747474 - 75967867 on Build GRCh38 | Loss |
|

Genomic Map